Product Details

SNP ID
rs12522205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:38258831 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCGGACCCGGCGCGGTGTCGCTCC[C/G]AGCGGCCATCCGAAAACCAGGTAAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EGFLAM PubMed Links

Gene Details

Gene
EGFLAM
Gene Name
EGF like, fibronectin type III and laminin G domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205301.1 423 Missense Mutation CCA,CGA P26R NP_001192230.1
NM_152403.3 423 Missense Mutation CCA,CGA P26R NP_689616.2
NM_182798.2 423 Intron NP_877950.1
NM_182801.2 423 Intron NP_877953.1

View Full Product Details