Product Details

SNP ID
rs755123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17325075 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCAGCCAGTGAGGATGTGGCCCC[C/T]GACGAGAGGAATTTGCCCTGCAGTG
Phenotype
MIM: 610216 MIM: 608536
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANO8 PubMed Links

Gene Details

Gene
ANO8
Gene Name
anoctamin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020959.2 3132 Silent Mutation TCA,TCG S991S NP_066010.1
XM_017027048.1 3132 Silent Mutation TCA,TCG S991S XP_016882537.1
Gene
DDA1
Gene Name
DET1 and DDB1 associated 1
There are no transcripts associated with this gene.

Gene
GTPBP3
Gene Name
GTP binding protein 3 (mitochondrial)
There are no transcripts associated with this gene.

View Full Product Details