Product Details

SNP ID
rs13273607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:26744049 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTCTTTCTTCCACTGACACAGAGC[A/T]TGTCAAAGGTCTAGAATAGCAGCTG
Phenotype
MIM: 104221
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ADRA1A PubMed Links

Gene Details

Gene
ADRA1A
Gene Name
adrenoceptor alpha 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000680.3 10573 Intron NP_000671.2
NM_001322502.1 10573 Intron NP_001309431.1
NM_001322503.1 10573 Intron NP_001309432.1
NM_001322504.1 10573 Intron NP_001309433.1
NM_033302.3 10573 Intron NP_150645.2
NM_033303.4 10573 Intron NP_150646.3
NM_033304.3 10573 Intron NP_150647.2
XM_006716292.3 10573 UTR 3 XP_006716355.1
XM_006716293.3 10573 Intron XP_006716356.1
XM_011544411.2 10573 Intron XP_011542713.1
XM_011544412.2 10573 Intron XP_011542714.1
XM_017013094.1 10573 UTR 3 XP_016868583.1
XM_017013095.1 10573 UTR 3 XP_016868584.1
XM_017013096.1 10573 Intron XP_016868585.1

View Full Product Details