Product Details

SNP ID
rs11629206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93923282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCACTGCGCCCGGCTGAATCTC[C/T]ACATATTTTTAAATGGGCATCTCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM181A PubMed Links

Gene Details

Gene
FAM181A
Gene Name
family with sequence similarity 181 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207071.1 Intron NP_001194000.1
NM_001207072.1 Intron NP_001194001.1
NM_001207073.1 Intron NP_001194002.1
NM_001207074.1 Intron NP_001194003.1
NM_138344.4 Intron NP_612353.3
XM_011537297.1 Intron XP_011535599.1
Gene
FAM181A-AS1
Gene Name
FAM181A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details