Product Details

SNP ID
rs9387383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116101684 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATTCCAGTCCTACCAACAGTCTGC[A/G]AAGTTTGGGTCCTCACTGTGCCCTG
Phenotype
MIM: 606573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRK PubMed Links

Gene Details

Gene
FRK
Gene Name
fyn related Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002031.2 Intron NP_002022.1
XM_005266880.4 Intron XP_005266937.1
XM_005266881.2 Intron XP_005266938.1
XM_005266882.4 Intron XP_005266939.1
XM_011535653.2 Intron XP_011533955.1
XM_011535654.2 Intron XP_011533956.1
XM_011535655.2 Intron XP_011533957.1
XM_011535656.2 Intron XP_011533958.1
XM_017010645.1 Intron XP_016866134.1
Gene
NT5DC1
Gene Name
5'-nucleotidase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152729.2 Intron NP_689942.2
XM_006715378.3 Intron XP_006715441.1

View Full Product Details