Product Details

SNP ID
rs3732966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9974056 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGATTAAAAGCGAATCAACTTA[C/T]AAACAAACAGAAAACTCACCCTAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMC3 PubMed Links

Gene Details

Gene
EMC3
Gene Name
ER membrane protein complex subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018447.3 Intron NP_060917.1
XM_005265321.3 Intron XP_005265378.1

View Full Product Details