Product Details

SNP ID
rs6473911
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:54136362 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAAAGATGCTAACAGAGTGTGTGGA[C/G]CTTTGAGCAACTAGGACATGCTTGA
Phenotype
MIM: 611828
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRPL15 PubMed Links

Gene Details

Gene
MRPL15
Gene Name
mitochondrial ribosomal protein L15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014175.3 Intron NP_054894.1
XM_017013362.1 Intron XP_016868851.1

View Full Product Details