Product Details

SNP ID
rs11708107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:132320154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAAGTCTTTAACATGCTAATAC[A/G]CACTGTGGCTTTCAAGGAAAGGATC
Phenotype
MIM: 171790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACPP PubMed Links

Gene Details

Gene
ACPP
Gene Name
acid phosphatase, prostate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099.4 Intron NP_001090.2
NM_001134194.1 Intron NP_001127666.1
NM_001292037.1 Intron NP_001278966.1
XM_011512946.1 Intron XP_011511248.1
XM_011512947.2 Intron XP_011511249.1

View Full Product Details