Product Details

SNP ID
rs12962570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:3415543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCAGCACCCCCTTTAGAGCCC[A/G]CAGACCAGGACCCTCCAGCTCCAGC
Phenotype
MIM: 602630
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TGIF1 PubMed Links

Gene Details

Gene
TGIF1
Gene Name
TGFB induced factor homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278682.1 Intron NP_001265611.1
NM_001278684.1 Intron NP_001265613.1
NM_001278686.1 Intron NP_001265615.1
NM_003244.3 Intron NP_003235.1
NM_170695.3 Intron NP_733796.2
NM_173207.2 Intron NP_775299.1
NM_173208.2 Intron NP_775300.1
NM_173209.2 Intron NP_775301.1
NM_173210.2 Intron NP_775302.1
NM_173211.1 Intron NP_775303.1
NM_174886.2 Intron NP_777480.1
XM_011525735.2 Intron XP_011524037.1
XM_017025958.1 Intron XP_016881447.1
XM_017025959.1 Intron XP_016881448.1

View Full Product Details