Product Details
- SNP ID
-
rs340832
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:3067948 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAGGCAGTTTCCACAATGCCAACT[C/G]TCTGGGTTGTGGTTTGAAAGAGATC
- Phenotype
-
MIM: 607280
MIM: 147851
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CNTN4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77247039] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CNTN4
- Gene Name
- contactin 4
There are no transcripts associated with this gene.
- Gene
- CNTN4-AS1
- Gene Name
- CNTN4 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- IL5RA
- Gene Name
- interleukin 5 receptor subunit alpha
View Full Product Details