Product Details

SNP ID
rs11998699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:58494221 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGTAAGAGGAATATGAGAAACAT[A/G]GTGTGGCAGTGCAGGGTGGTAGTTG
Phenotype
MIM: 118455
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP7A1 PubMed Links

Gene Details

Gene
CYP7A1
Gene Name
cytochrome P450 family 7 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000780.3 Intron NP_000771.2

View Full Product Details