Product Details

SNP ID
rs12459549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16549946 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCTTGAAAACAGGTGACAATTGAA[A/G]AACATTATAATATTTTCAGGTCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHERP PubMed Links

Gene Details

Gene
CHERP
Gene Name
calcium homeostasis endoplasmic reticulum protein
There are no transcripts associated with this gene.

Gene
SLC35E1
Gene Name
solute carrier family 35 member E1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024881.4 4984 UTR 3 NP_079157.3

View Full Product Details