Product Details

SNP ID
rs3826828
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:48333416 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGGAGCCAGTTCTCCACCTCCGACC[G/T]GAGCCACCTGGAGGTCTCTGCAAGG
Phenotype
MIM: 602335
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EMP3 PubMed Links

Gene Details

Gene
EMP3
Gene Name
epithelial membrane protein 3
There are no transcripts associated with this gene.

Gene
TMEM143
Gene Name
transmembrane protein 143
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303538.1 1395 Silent Mutation AGG,CGG R360R NP_001290467.1
NM_001303539.1 1395 Silent Mutation AGG,CGG R330R NP_001290468.1
NM_001303540.1 1395 Silent Mutation AGG,CGG R295R NP_001290469.1
NM_018273.3 1395 Silent Mutation AGG,CGG R395R NP_060743.2

View Full Product Details