Product Details
- SNP ID
-
rs12610706
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:49852927 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCATCTCAGATGCTGGATGTCCGGT[A/G]TGGTGCCTGAAGCCCCCGGGGCAAC
- Phenotype
-
MIM: 605610
MIM: 610195
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR4749
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77941735] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR4749
- Gene Name
- microRNA 4749
There are no transcripts associated with this gene.
- Gene
- PNKP
- Gene Name
- polynucleotide kinase 3'-phosphatase
There are no transcripts associated with this gene.
- Gene
- PTOV1
- Gene Name
- prostate tumor overexpressed 1
- Gene
- PTOV1-AS1
- Gene Name
- PTOV1 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- PTOV1-AS2
- Gene Name
- PTOV1 antisense RNA 2
There are no transcripts associated with this gene.
View Full Product Details