Product Details

SNP ID
rs965054
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:20536708 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTGGAGTTCAAGAATTTTCATTTC[A/T]GGCTGGGCATGGTGGCTCATGCCTG
Phenotype
MIM: 611259
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CDKAL1 PubMed Links

Gene Details

Gene
CDKAL1
Gene Name
CDK5 regulatory subunit associated protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017774.3 Intron NP_060244.2
XM_011514718.1 Intron XP_011513020.1
XM_011514719.2 Intron XP_011513021.1
XM_017010986.1 Intron XP_016866475.1
XM_017010987.1 Intron XP_016866476.1

View Full Product Details