Product Details
- SNP ID
-
rs922810
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:22580800 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGGCTTGCTTTTTCTGTTCTGGAGC[C/T]AGGGATCTGCTGCTGCCTGGCGTGC
- Phenotype
-
MIM: 609722
MIM: 610795
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PDLIM2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76598232] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PDLIM2
- Gene Name
- PDZ and LIM domain 2
- Gene
- SORBS3
- Gene Name
- sorbin and SH3 domain containing 3
There are no transcripts associated with this gene.
View Full Product Details