Product Details

SNP ID
rs4955393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:38287473 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGAATTTTTGTTTATGATATGA[C/T]GAATCTAATTTTAATTTATTTTTCC
Phenotype
MIM: 604047 MIM: 604048
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC22A13 PubMed Links

Gene Details

Gene
SLC22A13
Gene Name
solute carrier family 22 member 13
There are no transcripts associated with this gene.

Gene
SLC22A14
Gene Name
solute carrier family 22 member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320033.1 Intron NP_001306962.1
NM_004803.4 Intron NP_004794.2
XM_005265585.4 Intron XP_005265642.1
XM_006713416.3 Intron XP_006713479.2
XM_006713417.3 Intron XP_006713480.2
XM_006713418.3 Intron XP_006713481.2
XM_011534245.2 Intron XP_011532547.1
XM_011534247.2 Intron XP_011532549.1

View Full Product Details