Product Details

SNP ID
rs12133576
Assay Type
Functionally tested
NCBI dbSNP Submissions
58
Location
Chr.1:93350843 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATATTTTTCTGTTTTGGAAAG[A/G]TGAGATCCTTAGAAATAGGGCCATG
Phenotype
MIM: 601482
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC18-AS1 PubMed Links

Gene Details

Gene
CCDC18-AS1
Gene Name
CCDC18 antisense RNA 1
There are no transcripts associated with this gene.

Gene
DR1
Gene Name
down-regulator of transcription 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001938.2 Intron NP_001929.1

View Full Product Details