Product Details
- SNP ID
-
rs219641
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:26467463 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTAAGGAAATCTACAGAAGGGAATC[C/T]GAACTTCATTTCCCACTTTATTTTT
- Phenotype
-
MIM: 616020
MIM: 616021
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CYYR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76236577] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CYYR1
- Gene Name
- cysteine and tyrosine rich 1
- Gene
- CYYR1-AS1
- Gene Name
- cysteine and tyrosine rich 1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details