Product Details

SNP ID
rs240985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:111263428 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCTATTTCCCAAACTAATTTATT[C/T]AGTAAATTTTATTACAAACTACTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFSD4B PubMed Links

Gene Details

Gene
MFSD4B
Gene Name
major facilitator superfamily domain containing 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153369.2 Intron NP_699200.2
XM_017011483.1 Intron XP_016866972.1

View Full Product Details