Product Details

SNP ID
rs694383
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:182599069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTGGGCTCTGCCGCAGGAGTT[C/G]CCATCAGGAAATGTTTGTTCAGTGA
Phenotype
MIM: 602514 MIM: 180435
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RGS16 PubMed Links

Gene Details

Gene
RGS16
Gene Name
regulator of G-protein signaling 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002928.3 1986 UTR 3 NP_002919.3
Gene
RNASEL
Gene Name
ribonuclease L
There are no transcripts associated with this gene.

View Full Product Details