Product Details

SNP ID
rs659046
Assay Type
Validated
NCBI dbSNP Submissions
71
Location
Chr.1:20721232 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
ACCCCTCCTTCACTCTCAGATACAT[A/G]TGAGAAACAGCCCTGCTCTGTCAGC
Phenotype
MIM: 605037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF17 PubMed Links

Gene Details

Gene
KIF17
Gene Name
kinesin family member 17
There are no transcripts associated with this gene.

Gene
SH2D5
Gene Name
SH2 domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001103160.1 1912 UTR 3 NP_001096630.1
NM_001103161.1 1912 UTR 3 NP_001096631.1
XM_011541459.1 1912 UTR 3 XP_011539761.1
XM_011541460.1 1912 UTR 3 XP_011539762.1
XM_011541461.2 1912 UTR 3 XP_011539763.1
XM_011541462.1 1912 UTR 3 XP_011539764.1

View Full Product Details