Product Details

SNP ID
rs17408061
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:20743745 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACACCAGAAATCACAAATCAATA[C/T]ATTTCTACACTTCCCCTTCAGTCAA
Phenotype
MIM: 616072
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HP1BP3 PubMed Links

Gene Details

Gene
HP1BP3
Gene Name
heterochromatin protein 1 binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016287.3 3290 UTR 3 NP_057371.2
XM_005245875.4 3290 UTR 3 XP_005245932.1
XM_005245876.3 3290 UTR 3 XP_005245933.1
XM_005245877.4 3290 UTR 3 XP_005245934.1
XM_005245878.4 3290 UTR 3 XP_005245935.1
XM_005245879.4 3290 UTR 3 XP_005245936.1
XM_011541532.1 3290 UTR 3 XP_011539834.1
XM_011541533.1 3290 UTR 3 XP_011539835.1
XM_011541534.2 3290 UTR 3 XP_011539836.1
XM_017001393.1 3290 UTR 3 XP_016856882.1
XM_017001394.1 3290 UTR 3 XP_016856883.1

View Full Product Details