Product Details

SNP ID
rs1142887
Assay Type
Functionally Tested
NCBI dbSNP Submissions
14
Location
Chr.1:89186411 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTCCAACTTCTTCTCCATTTG[G/A]GCCATGTATTCCTGGAAGCTTCTCT
Phenotype
MIM: 612466
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
GBP4 PubMed Links

Gene Details

Gene
GBP4
Gene Name
guanylate binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052941.4 1745 Silent Mutation GCC,GCT A543A NP_443173.2

View Full Product Details