Product Details

SNP ID
rs623134
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:89185167 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTGAATGAAACTGCTATAACAC[A/T]TATACTTACAAAATGAGCAACAGTG
Phenotype
MIM: 612466 MIM: 612468
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GBP4 PubMed Links

Gene Details

Gene
GBP4
Gene Name
guanylate binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052941.4 2126 UTR 3 NP_443173.2
Gene
GBP7
Gene Name
guanylate binding protein 7
There are no transcripts associated with this gene.

View Full Product Details