Product Details

SNP ID
rs8125955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3039311 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGTATCCTTAGGATATTATTCTC[A/C]TCCTCATGGTTGAAGCTGGGTCACT
Phenotype
MIM: 602352 MIM: 611988 MIM: 176884
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GNRH2 PubMed Links

Gene Details

Gene
GNRH2
Gene Name
gonadotropin releasing hormone 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001310220.1 251 Intron NP_001297149.1
NM_001501.1 251 Intron NP_001492.1
NM_178331.1 251 Intron NP_847901.1
NM_178332.1 251 Intron NP_847902.1
XM_017027823.1 251 UTR 5 XP_016883312.1
Gene
MRPS26
Gene Name
mitochondrial ribosomal protein S26
There are no transcripts associated with this gene.

Gene
PTPRA
Gene Name
protein tyrosine phosphatase, receptor type A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002836.3 251 Intron NP_002827.1
NM_080840.2 251 Intron NP_543030.1
NM_080841.2 251 Intron NP_543031.1

View Full Product Details