Product Details
- SNP ID
-
rs3010332
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
40
- Location
-
Chr.1:59132560 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTCCTGAAGCACAGGCAGGAGGTC[C/T]CCGTCACAAGCTATTGATTTGGTGT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HSD52
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79533696] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HSD52
- Gene Name
- uncharacterized LOC729467
There are no transcripts associated with this gene.
View Full Product Details