Product Details

SNP ID
rs17174597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10160092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTGCCACCTTGTTTCAGAAACT[C/T]AGCTTTTGAAACTTTTTGTCTTAAA
Phenotype
MIM: 602601
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OLR1 PubMed Links

Gene Details

Gene
OLR1
Gene Name
oxidized low density lipoprotein receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172632.1 Intron NP_001166103.1
NM_001172633.1 Intron NP_001166104.1
NM_002543.3 Intron NP_002534.1

View Full Product Details