Product Details

SNP ID
rs2371455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56450109 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAATTATTAAGCATTTGAATAGAA[G/T]GAATCAATTTCTGGGGTATGAGGGA
Phenotype
MIM: 154050 MIM: 603887
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIP PubMed Links

Gene Details

Gene
MIP
Gene Name
major intrinsic protein of lens fiber
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012064.3 1742 UTR 3 NP_036196.1
XM_011538354.1 1742 UTR 3 XP_011536656.1
XM_017019306.1 1742 UTR 3 XP_016874795.1
Gene
TIMELESS
Gene Name
timeless circadian clock
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011538939.2 1742 Intron XP_011537241.1

View Full Product Details