Product Details

SNP ID
rs11649081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21303291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGCCTAGAGTCAGGAAAAAGCC[C/T]TTTCAAGTCAAGCTGAGAACTTTAA
Phenotype
MIM: 123740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRYM PubMed Links

Gene Details

Gene
CRYM
Gene Name
crystallin mu
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001888.4 Intron NP_001879.1
Gene
CRYM-AS1
Gene Name
CRYM antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details