Product Details

SNP ID
rs34164618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55578710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGCGGTGCTGGCGGAGGTAGGA[G/T]GCGAGGCGGAAGGCCAGGCCGCAGT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF579 PubMed Links

Gene Details

Gene
ZNF579
Gene Name
zinc finger protein 579
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152600.2 983 Silent Mutation GCA,GCC A310A NP_689813.2
XM_017026409.1 983 Silent Mutation GCA,GCC A310A XP_016881898.1
XM_017026410.1 983 Silent Mutation GCA,GCC A310A XP_016881899.1
XM_017026411.1 983 Silent Mutation GCA,GCC A310A XP_016881900.1

View Full Product Details