Product Details

SNP ID
rs12609925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39973146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCATGAAATATTTAGCCTTACTA[G/T]GTGTGGTATATGCTGATGTTTTCTA
Phenotype
MIM: 602707
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PSMC4 PubMed Links

Gene Details

Gene
PSMC4
Gene Name
proteasome 26S subunit, ATPase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006503.3 Intron NP_006494.1
NM_153001.2 Intron NP_694546.1

View Full Product Details