Product Details

SNP ID
rs770375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:89423587 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATTTCTAATTATAGTTCAGTAA[C/T]GATAAACACCGTCCAACTTAGTTAG
Phenotype
MIM: 614784
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105369889 PubMed Links

Gene Details

Gene
LOC105369889
Gene Name
uncharacterized LOC105369889
There are no transcripts associated with this gene.

Gene
POC1B
Gene Name
POC1 centriolar protein B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199777.1 Intron NP_001186706.1
NM_172240.2 Intron NP_758440.1

View Full Product Details