Product Details

SNP ID
rs2069408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55970537 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATGTAATGTGCATTTATCCCCCC[A/G]GTGCATTACCTTACAATTGTCCGTA
Phenotype
MIM: 116953 MIM: 155550 MIM: 179514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK2 PubMed Links

Gene Details

Gene
CDK2
Gene Name
cyclin dependent kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290230.1 Intron NP_001277159.1
NM_001798.4 Intron NP_001789.2
NM_052827.3 Intron NP_439892.2
XM_011537732.1 Intron XP_011536034.1
Gene
PMEL
Gene Name
premelanosome protein
There are no transcripts associated with this gene.

Gene
RAB5B
Gene Name
RAB5B, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details