Product Details

SNP ID
rs1976270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:85043131 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGGTTTTTTGTTTATTTTTATAA[A/G]GAGAGATGTTCTGAATATTTATTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRIQ1 PubMed Links

Gene Details

Gene
LRRIQ1
Gene Name
leucine rich repeats and IQ motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079910.1 Intron NP_001073379.1
XM_011538817.2 Intron XP_011537119.1
XM_011538818.2 Intron XP_011537120.1
XM_011538819.2 Intron XP_011537121.1
XM_011538823.2 Intron XP_011537125.1
XM_011538824.2 Intron XP_011537126.1
XM_011538825.2 Intron XP_011537127.1
XM_017020019.1 Intron XP_016875508.1
Gene
TSPAN19
Gene Name
tetraspanin 19
There are no transcripts associated with this gene.

View Full Product Details