Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286250.1 | 619 | Intron | NP_001273179.1 | ||
NM_001286251.1 | 619 | Intron | NP_001273180.1 | ||
NM_001286252.1 | 619 | Intron | NP_001273181.1 | ||
NM_001286254.2 | 619 | Intron | NP_001273183.1 | ||
NM_001323252.1 | 619 | Intron | NP_001310181.1 | ||
NM_001323253.1 | 619 | Intron | NP_001310182.1 | ||
NM_001323254.1 | 619 | Intron | NP_001310183.1 | ||
NM_001323255.1 | 619 | Intron | NP_001310184.1 | ||
NM_001323256.1 | 619 | Intron | NP_001310185.1 | ||
NM_001323257.1 | 619 | Intron | NP_001310186.1 | ||
NM_001323258.1 | 619 | UTR 5 | NP_001310187.1 | ||
NM_001323259.1 | 619 | Intron | NP_001310188.1 | ||
NM_001323260.1 | 619 | Intron | NP_001310189.1 | ||
NM_001323261.1 | 619 | Intron | NP_001310190.1 | ||
NM_001323262.1 | 619 | Intron | NP_001310191.1 | ||
NM_001323263.1 | 619 | Intron | NP_001310192.1 | ||
NM_001323264.1 | 619 | Intron | NP_001310193.1 | ||
NM_052904.3 | 619 | Intron | NP_443136.2 | ||
XM_005266813.4 | 619 | Intron | XP_005266870.1 | ||
XM_017010228.1 | 619 | Intron | XP_016865717.1 | ||
XM_017010229.1 | 619 | Intron | XP_016865718.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014165.3 | 619 | Intron | NP_054884.1 |