Product Details

SNP ID
rs10759408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:110306678 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATGAGCTCATCTGCTAGTGATAA[A/C]TTTTTACTATTGTTGTGTTATTATC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TXNDC8 PubMed Links

Gene Details

Gene
TXNDC8
Gene Name
thioredoxin domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003936.3 Intron NP_001003936.1
NM_001286946.1 Intron NP_001273875.1
NM_001286947.1 Intron NP_001273876.1
XM_011518463.2 Intron XP_011516765.1
XM_017014579.1 Intron XP_016870068.1
XM_017014580.1 Intron XP_016870069.1
XM_017014581.1 Intron XP_016870070.1
XM_017014582.1 Intron XP_016870071.1
XM_017014583.1 Intron XP_016870072.1
XM_017014584.1 Intron XP_016870073.1

View Full Product Details