Product Details

SNP ID
rs2302905
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:31405874 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAATCTCCTCTGATTACTTACAGT[A/G]TCACACACTGAATTGCACAGATAGG
Phenotype
MIM: 608828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DROSHA PubMed Links

Gene Details

Gene
DROSHA
Gene Name
drosha ribonuclease III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100412.1 Intron NP_001093882.1
NM_013235.4 Intron NP_037367.3
XM_005248291.3 Intron XP_005248348.1
XM_005248292.3 Intron XP_005248349.1
XM_005248293.3 Intron XP_005248350.1
XM_005248294.3 Intron XP_005248351.1
XM_011514033.2 Intron XP_011512335.1
XM_017009399.1 Intron XP_016864888.1
XM_017009400.1 Intron XP_016864889.1
XM_017009401.1 Intron XP_016864890.1
XM_017009402.1 Intron XP_016864891.1

View Full Product Details