Product Details

SNP ID
rs8111400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37932114 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCTGCATTGCCTCTGTGAGCTAG[C/T]GCCTCCTCTGCACGGGCCAAGGAGC
Phenotype
MIM: 616655
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SIPA1L3 PubMed Links

Gene Details

Gene
SIPA1L3
Gene Name
signal induced proliferation associated 1 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015073.2 355 Intron NP_055888.1
XM_005258671.4 355 Intron XP_005258728.1
XM_011526657.2 355 UTR 5 XP_011524959.1
XM_017026517.1 355 UTR 5 XP_016882006.1
XM_017026518.1 355 Intron XP_016882007.1

View Full Product Details