Product Details

SNP ID
rs13239774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:65394154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCATAAGTGGTGCCAAGACCAAT[A/G]TAATGTCTTTCCTCTATATTGTTCC
Phenotype
MIM: 603974
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF92 PubMed Links

Gene Details

Gene
ZNF92
Gene Name
zinc finger protein 92
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287532.1 Intron NP_001274461.1
NM_001287533.1 Intron NP_001274462.1
NM_001287534.1 Intron NP_001274463.1
NM_007139.3 Intron NP_009070.2
NM_152626.3 Intron NP_689839.1

View Full Product Details