Product Details

SNP ID
rs2502364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38543087 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCACAAGGGCAGTAAACACCCAC[A/G]GGTGAGGTATGTCCACTCTCCACAC
Phenotype
MIM: 610786
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SRCIN1 PubMed Links

Gene Details

Gene
SRCIN1
Gene Name
SRC kinase signaling inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025248.2 3859 Intron NP_079524.2
XM_017025169.1 3859 Intron XP_016880658.1
XM_017025170.1 3859 Intron XP_016880659.1
XM_017025171.1 3859 Intron XP_016880660.1
XM_017025172.1 3859 Intron XP_016880661.1
XM_017025173.1 3859 UTR 3 XP_016880662.1
XM_017025174.1 3859 UTR 3 XP_016880663.1

View Full Product Details