Product Details

SNP ID
rs1078543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10765628 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGCTGCTCTTCTGCTGGCGCGC[A/T]CTTCTTCTTGGCCACTCGCTCCTGG
Phenotype
MIM: 606505
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC102723313 PubMed Links

Gene Details

Gene
LOC102723313
Gene Name
uncharacterized LOC102723313
There are no transcripts associated with this gene.

Gene
PINX1
Gene Name
PIN2/TERF1 interacting, telomerase inhibitor 1
There are no transcripts associated with this gene.

View Full Product Details