Product Details

SNP ID
rs71417335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105042423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTCCCTCAGGCTGATGAAGAA[A/C]AGGTAAGCAAGAGAGAAGAACAGCA
Phenotype
MIM: 611975
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC102724691 PubMed Links

Gene Details

Gene
LOC102724691
Gene Name
uncharacterized LOC102724691
There are no transcripts associated with this gene.

Gene
MRPS9
Gene Name
mitochondrial ribosomal protein S9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182640.2 Intron NP_872578.1
XM_011511644.2 Intron XP_011509946.1

View Full Product Details