Product Details

SNP ID
rs6545835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:60885417 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATTTATTTATTTTACCCTGGCAC[G/T]TGCAAGAGTTTTTTCAAGACTTACC
Phenotype
MIM: 164910
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC01185 PubMed Links

Gene Details

Gene
LINC01185
Gene Name
long intergenic non-protein coding RNA 1185
There are no transcripts associated with this gene.

Gene
REL
Gene Name
REL proto-oncogene, NF-kB subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291746.1 Intron NP_001278675.1
NM_002908.3 Intron NP_002899.1
XM_011533010.2 Intron XP_011531312.1
XM_017004627.1 Intron XP_016860116.1

View Full Product Details