Product Details

SNP ID
rs11955293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157788019 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACCAGACGAAAACAGACAGAAGAA[C/T]TTTACCACAATAAATTTTTAGGTTC
Phenotype
MIM: 607265
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLINT1 PubMed Links

Gene Details

Gene
CLINT1
Gene Name
clathrin interactor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195555.1 Intron NP_001182484.1
NM_001195556.1 Intron NP_001182485.1
NM_014666.3 Intron NP_055481.1
XM_011534701.2 Intron XP_011533003.1
XM_017010087.1 Intron XP_016865576.1
XM_017010088.1 Intron XP_016865577.1

View Full Product Details