Product Details

SNP ID
rs10040588
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149862144 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAAGGAGGGCAGTCAGGGTGAGG[C/G]AGGGACAAGATGCCATGTCCTGTGA
Phenotype
MIM: 180071 MIM: 608886
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PDE6A PubMed Links

Gene Details

Gene
PDE6A
Gene Name
phosphodiesterase 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000440.2 Intron NP_000431.2
XM_011537650.2 Intron XP_011535952.1
XM_011537651.2 Intron XP_011535953.1
XM_011537652.1 Intron XP_011535954.1
XM_011537653.2 Intron XP_011535955.1
XM_011537654.2 Intron XP_011535956.1
XM_017009572.1 Intron XP_016865061.1
Gene
PPARGC1B
Gene Name
PPARG coactivator 1 beta
There are no transcripts associated with this gene.

View Full Product Details