Product Details
- SNP ID
-
rs184401
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:57472889 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGCTGTTCCGGCCCTTGGGCTGT[C/T]TGGGAAAAGCCAGCAGTCAGAGTGC
- Phenotype
-
MIM: 608333
MIM: 180663
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DOK4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114023919] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DOK4
- Gene Name
- docking protein 4
- Gene
- POLR2C
- Gene Name
- RNA polymerase II subunit C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032940.2 |
1878 |
Intron |
|
|
NP_116558.1 |
View Full Product Details