Product Details

SNP ID
rs166849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:199914143 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAAATTATTGGAGTATCCTAGTC[A/G]TCTGTTCTTCCATGTTTCCTCTGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf69 PubMed Links

Gene Details

Gene
C2orf69
Gene Name
chromosome 2 open reading frame 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153689.5 Intron NP_710156.3

View Full Product Details