Product Details

SNP ID
rs11216313
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117200646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGAGGAGGAAGGGGAAGGATG[A/G]TGGTGCTGGGTGGGAGTGGGGATGG
Phenotype
MIM: 604872 MIM: 600818
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100652768 PubMed Links

Gene Details

Gene
LOC100652768
Gene Name
uncharacterized LOC100652768
There are no transcripts associated with this gene.

Gene
PCSK7
Gene Name
proprotein convertase subtilisin/kexin type 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004716.3 Intron NP_004707.2
Gene
SIDT2
Gene Name
SID1 transmembrane family member 2
There are no transcripts associated with this gene.

Gene
TAGLN
Gene Name
transgelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001522.1 Intron NP_001001522.1
NM_003186.3 Intron NP_003177.2

View Full Product Details