Product Details

SNP ID
rs3825878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:84631524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGCTCGGGCTTCTTGCGTCCC[C/T]GCTAAGAACATGTCACGGGGCCGAA
Phenotype
MIM: 610417 MIM: 616144
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCAND2P PubMed Links

Gene Details

Gene
SCAND2P
Gene Name
SCAN domain containing 2 pseudogene
There are no transcripts associated with this gene.

Gene
WDR73
Gene Name
WD repeat domain 73
There are no transcripts associated with this gene.

Gene
ZSCAN2
Gene Name
zinc finger and SCAN domain containing 2
There are no transcripts associated with this gene.

View Full Product Details